6-Chloronicotinonitrile ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Chloro-5-cyanopyridine, 6-Chloro-3-pyridinecarbonitrile, 2-Chloropyridine-5-carbonitrile
A13291.06 A13291.03 A13291.14
A13291.06EA 1550 SEK
A13291.06 A13291.03 A13291.14
6-Chloronicotinonitrile ≥98%
6-Chloronicotinonitrile
Purity: 98% CAS No.: 33252-28-7 Molecular Formula: C6H3ClN2 Formula Weight: 138.56 Melting Point: 116-119° Boiling Point: 105-107°/1mm UN No.: 3439 MDL No.: MFCD00084941
Formel: C₆H₃ClN₂
MW: 138,56 g/mol
Kokpunkt: 105…107 °C (1 mmHg)
Smältpunkt: 116…119 °C
Storage Temperature: Omgivande
MDL Number: MFCD00084941
CAS nummer: 33252-28-7
UN: 3439
ADR: 6.1,III