N-metyl-N-(trimetylsilyl) trifluoracetamid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
A13141.06 A13141.14 A13141.22
A13141.06EA 1750 SEK
A13141.06 A13141.14 A13141.22
N-metyl-N-(trimetylsilyl) trifluoracetamid 97%
N-metyl-N-(trimetylsilyl) trifluoracetamid
Purity: 97% CAS No.: 24589-78-4 Molecular Formula: C6H12F3NOSi Formula Weight: 199.25 Flash Point: 25°(77°F) Refractive Index: 1.3800 Boiling Point: 130-132° Beilstein Reference: 1941550 UN No.: 1993 MDL No.: MFCD00000411
Formel: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Kokpunkt: 130…132 °C
Densitet: 1.076
Flampunkt: 25 °C (77 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000411
CAS nummer: 24589-78-4
EINECS: 246-331-6
ADR: 3 III