Anilin HCl ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride
A13024.0E A13024.22 A13024.36
A13024.0EEA 5775 SEK
A13024.0E A13024.22 A13024.36
Anilin HCl ≥99%
Anilin HCl
Purity: 99% CAS No.: 142-04-1 Molecular Formula: C6H7N.HCl Formula Weight: 129.59 Flash Point: 193°(379°F) Melting Point: 196-200° Boiling Point: 245° Beilstein Reference: 3593823 Merck Reference: 14,659 UN No.: 1548 MDL No.: MFCD00012958
Formel: C₆H₈ClN
Kokpunkt: 245 °C
Smältpunkt: 196…200 °C
Densitet: 1,221
Flampunkt: 193 °C (379 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00012958
CAS nummer: 142-04-1
EINECS: 205-519-8
UN: 1548
ADR: 6.1,III
Merck Index: 14,659