Iodotrimethylsilane ≥97% stabilised
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Trimethyliodosilane, Trimethylsilyl iodide
Stabilization:
Stabilised with copper
A12902.06
A12902.14
A12902.22
A12902.30
A12902.06EA
835
SEK
A12902.06
A12902.14
A12902.22
A12902.30
Iodotrimethylsilane ≥97% stabilised
Iodotrimethylsilane
Stabilized with copper
Purity: 97%
CAS No.: 16029-98-4
Molecular Formula: C3H9ISi
Formula Weight: 200.09
Storage Temperature: -30°C to -10°C
Flash Point: -2°(28°F)
Refractive Index: 1.4760
Boiling Point: 105-106°
Beilstein Reference: 1731136
UN No.: 2924
MDL No.: MFCD00001028
Formel:
C₃H₉ISi Kokpunkt: 105…106 °C Smältpunkt: < 0 °C Densitet: 1,440 Flampunkt: –2 °C (28 °F) Storage Temperature: Frys |
MDL Number:
MFCD00001028 CAS nummer: 16029-98-4 EINECS: 240-171-0 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...