Tetrabutylammonium hydroxide 40% (w/w) i metanol
Leverantör: Thermo Fisher Scientific
Fara
A12626.0F A12626.AE A12626.AP
A12626.0FEA 16200 SEK
A12626.0F A12626.AE A12626.AP
Tetrabutylammonium hydroxide 40% (w/w) i metanol
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5 Molecular Formula: [CH3(CH2)3]4NOH Formula Weight: 259.48 Flash Point: 18°(64°F) Refractive Index: 1.400 Odour: Alcohol-like Beilstein Reference: 3571228 UN No.: 2924 MDL No.: MFCD00009425
Formel: C₁₆H₃₇NO
MW: 259.48 g/mol
Smältpunkt: 29.85
Densitet: 0.910
Flampunkt: 18 °C (64 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3 II