Tris(dimethylamino)phosphine ≥97%
Leverantör: Thermo Fisher Scientific
Fara
A12571.06 A12571.09 A12571.14 A12571.18 A12571.30
A12571.06EA 1050 SEK
A12571.06 A12571.09 A12571.14 A12571.18 A12571.30
Tris(dimethylamino)phosphine ≥97%
Tris(dimethylamino)phosphine
Purity: 97% CAS No.: 1608-26-0 Molecular Formula: [(CH3)2N]3P Formula Weight: 163.21 Flash Point: 37°(98°F) Refractive Index: 1.4650 Melting Point: -44° Boiling Point: 162-164° Beilstein Reference: 906778 UN No.: 1993 MDL No.: MFCD00008301
Formel: C₆H₁₈N₃P
Kokpunkt: 162…164 °C
Smältpunkt: –44 °C
Densitet: 0,898
Flampunkt: 37 °C (98 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008301
CAS nummer: 1608-26-0
EINECS: 216-534-4
ADR: 3,III