Phenoxyacetic acid ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Phenoxyacetic acid
A12541.0E A12541.22 A12541.36
A12541.0EEA 4300 SEK
A12541.0E A12541.22 A12541.36
Phenoxyacetic acid ≥99%
Phenoxyacetic acid
Purity: 99% CAS No.: 122-59-8 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 165°(329°F) Melting Point: 98-101° Boiling Point: 285° Beilstein Reference: 907949 Merck Reference: 14,7254 MDL No.: MFCD00004296
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 285 °C
Smältpunkt: 98…101 °C
Densitet: 1,36 g/cm³ (20 °C)
Flampunkt: 165 °C (329 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004296
CAS nummer: 122-59-8
EINECS: 204-556-7
Merck Index: 14,7254