Ethyl crotonate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester
A12529.0E A12529.22 A12529.36
A12529.0EEA 11650 SEK
A12529.0E A12529.22 A12529.36
Ethyl crotonate ≥98%
Ethyl crotonate
Purity: 98% CAS No.: 623-70-1 Molecular Formula: CH3CH=CHCO2CH2CH3 Formula Weight: 114.14 Flash Point: 2°(36°F) Boiling Point: 141-143° UN No.: 1862 MDL No.: MFCD00009289
Formel: C₆H₁₀O₂
Kokpunkt: 141…143 °C
Smältpunkt: –79 °C
Densitet: 0,918
Flampunkt: 2 °C (36 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009289
CAS nummer: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625