N-(Trimetylsilyl)imidazol ≥97%
Leverantör: Thermo Fisher Scientific
Fara
A12512.06 A12512.18 A12512.22 A12512.30
A12512.06EA 495 SEK
A12512.06 A12512.18 A12512.22 A12512.30
N-(Trimetylsilyl)imidazol ≥97%
N-(Trimetylsilyl)imidazol
Purity: 97% CAS No.: 18156-74-6 Molecular Formula: C6H12N2Si Formula Weight: 140.26 Flash Point: 18°(64°F) Refractive Index: 1.4765 Odour: Weak Melting Point: -42° Boiling Point: 212° Beilstein Reference: 606148 UN No.: 1993 MDL No.: MFCD00005280
Formel: C₆H₁₂N₂Si
Kokpunkt: 212 °C
Smältpunkt: –42 °C
Densitet: 0,956
Flampunkt: 18 °C (64 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005280
CAS nummer: 18156-74-6
EINECS: 242-040-3
ADR: 3,II