1-Bromo-4-propoxybenzene ≥98%
Leverantör: Thermo Fisher Scientific
A11462.06 A11462.14
A11462.06EA 1600 SEK
A11462.06 A11462.14
1-Bromo-4-propoxybenzene ≥98%
1-Bromo-4-propoxybenzene
Purity: 98% CAS No.: 39969-56-7 Molecular Formula: C9H11BrO Formula Weight: 215.10 Flash Point: 110°(230°F) Boiling Point: 238-240° MDL No.: MFCD00156160
Formel: C₉H₁₁BrO
Kokpunkt: 238…240 °C
Densitet: 1,356
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00156160
CAS nummer: 39969-56-7