tetra-Kaliumdifosfat, vattenfri ≥96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate
44269.A1 44269.A7
44269.A1EA 1600 SEK
44269.A1 44269.A7
tetra-Kaliumdifosfat, vattenfri ≥96%
tetra-Kaliumdifosfat
Purity: 96% min CAS No.: 7320-34-5 Molecular Formula: K4O7P2 Formula Weight: 330.34 Physical Form: Granular Density: 2.33 Appearance: White Melting Point: 1090° Boiling Point: dec. Merck Reference: 14,7663 MDL No.: MFCD00011393
Formel: K₄O₇P₂
Kokpunkt: Dec.
Smältpunkt: 1090 °C
Densitet: 2,33
Storage Temperature: Omgivande
MDL Number: MFCD00011393
CAS nummer: 7320-34-5
EINECS: 230-785-7
Merck Index: 14,7663