Methyl levulinate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Ketovaleric acid methyl ester, 4-Oxopentanoic acid methyl ester, Methyl 4-oxopentanoate, 4-Oxovaleric acid methyl ester, Methyl 4-oxovalerate, Levulinic acid methyl ester
43397.22 43397.14
43397.22EA 4625 SEK
43397.22 43397.14
Methyl levulinate ≥99%
Methyl levulinate
Purity: 99+% CAS No.: 624-45-3 Molecular Formula: CH3COCH2CH2CO2CH3 Formula Weight: 130.14 Flash Point: 72°(161.6°F) Physical Form: Liquid Density: 1.05 Odour: Pleasant Boiling Point: 191° MDL No.: MFCD00017499
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Kokpunkt: 191 °C
Smältpunkt: –24 °C
Densitet: 1,05
Flampunkt: 72 °C (161,6 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00017499
CAS nummer: 624-45-3
EINECS: 210-846-4