Salpetersyra standardised solution 0.1 N
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Nitric acid solution
35626.AP 35626.K2
35626.APEA 1100 SEK
35626.AP 35626.K2
Salpetersyra standardised solution 0.1 N
Salpetersyra
CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 87
Smältpunkt: -42
Densitet: 1.51
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610