Diammonium hexachloroiridate (≥43% Ir) ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Ammonium hexachloroiridate(IV)
12326.03 12326.06
12326.03EA 8150 SEK
12326.03 12326.06
Diammonium hexachloroiridate (≥43% Ir) ≥99%
Diammonium hexachloroiridate
Purity: 99% CAS No.: 16940-92-4 Molecular Formula: (NH4)2IrCl6 Formula Weight: 441.02 Physical Form: Crystalline powder Density: 2.86 Appearance: Red-black Melting Point: dec. MDL No.: MFCD00010881
Formel: H₈Cl₆IrN₂
MW: 441,01 g/mol
Smältpunkt: Dec.
Densitet: 2,86
Storage Temperature: Omgivande
MDL Number: MFCD00010881
CAS nummer: 16940-92-4
EINECS: 241-007-0