Dipotassium hexachloroiridate (≥39% Ir)
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Potassium hexachloroiridate(IV), Potassium hexachloroiridate (IV)
11888.03 11888.06 11888.14
11888.03EA 7925 SEK
11888.03 11888.06 11888.14
Dipotassium hexachloroiridate (≥39% Ir)
Dipotassium hexachloroiridate
CAS No.: 16920-56-2 Molecular Formula: K2IrCl6 Formula Weight: 483.12 Physical Form: Crystalline Sensitivity: Hygroscopic Odour: Odourless MDL No.: MFCD00011369
Formel: Cl₆IrK₂
MW: 483.13 g/mol
Smältpunkt: Dec.
Storage Temperature: Omgivande
MDL Number: MFCD00011369
CAS nummer: 16920-56-2
EINECS: 240-976-7