Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9%
Leverantör: Thermo Scientific
Fara
Synonyms: Diammonium hexachloropalladate
11042.04 11042.09
11042.04EA 4450 SEK
11042.04 11042.09
Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9%
Ammonium hexachloropalladate(IV)
Purity: 99.9% CAS No.: 19168-23-1 Molecular Formula: (NH4)2PdCl6 Formula Weight: 355.20 Physical Form: Crystalline Density: 2.42 Appearance: Red-orange Odour: Odourless UN No.: 3077 MDL No.: MFCD00015959
Formel: H₈Cl₆N₂Pd
MW: 355,21 g/mol
Densitet: 2,42
Storage Temperature: Omgivande
MDL Number: MFCD00015959
CAS nummer: 19168-23-1
EINECS: 242-854-9
UN: 3077
ADR: 9,III