3,3,3-Trifluoropropionic acid 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 3,3,3-Trifluoropropanoic acid
381070010 381070050 381071000
ACRO381070010EA 785 SEK
ACRO381070010 ACRO381070050 ACRO381071000
3,3,3-Trifluoropropionic acid 98%
3,3,3-Trifluoropropionic acid
Formel: C₃H₃F₃O₂
Kokpunkt: 146 °C (760 mmHg)
Densitet: 1,45 g/cm³
Flampunkt: 92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00153292
CAS nummer: 2516-99-6
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3320 to 1.3340 (20°C, 589 nm)
Titration with NaOH ≥97.5 %