2-Amino-5-bromopyrimidine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine
368580010 368580050
ACRO368580010EA 650 SEK
ACRO368580010 ACRO368580050
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine
Formel: C₄H₄BrN₃
MW: 174 g/mol
Smältpunkt: 239…244 °C
Flampunkt: >300 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012341
CAS nummer: 7752-82-1
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 230 to 244 °C
HPLC ≥96.0 %