1-Ethyl-3-methylimidazolium chloride 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride
354080050 354080250 354081000
ACRO354080050EA 500 SEK
ACRO354080050 ACRO354080250 ACRO354081000
1-Ethyl-3-methylimidazolium chloride 97%
1-Ethyl-3-methylimidazolium chloride
Formel: C₆H₁₁ClN₂
Smältpunkt: 77…79 °C
Densitet: ~ 1,1 g/cm³ (20 °C)
Flampunkt: 186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074843
CAS nummer: 65039-09-0
Order Now
Specifikation testresultat
Appearance White to yellow to orange Adhering powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %