3,4,5-Trimethoxybenzylamine 96%
Leverantör: Thermo Fisher Scientific
Fara
351170050 351170250
ACRO351170050EA 1075 SEK
ACRO351170050 ACRO351170250
3,4,5-Trimethoxybenzylamine 96%
3,4,5-Trimethoxybenzylamine
Formel: C₁₀H₁₅NO₃
Kokpunkt: 106 °C (0,1 mmHg)
Densitet: 1,155 g/cm³
Flampunkt: >110 °C
MDL Number: MFCD00017150
CAS nummer: 18638-99-8
EINECS: 242-468-0
UN: 2735
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5510 (20°C, 589 nm)
GC ≥95 %