tert-Butyl chloroacetate 97%
Leverantör: Thermo Scientific
Fara
347230250 347231000 347235000
ACRO347230250EA 470 SEK
ACRO347230250 ACRO347231000 ACRO347235000
tert-Butyl chloroacetate 97%
tert-Butyl chloroacetate
Formel: C₆H₁₁ClO₂
MW: 150,6 g/mol
Kokpunkt: 48…49 °C (11 mmHg)
Densitet: 1,053 g/cm³
Flampunkt: 46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000930
CAS nummer: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 13,01562
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥96.0 %