3-Cyanophenyl isocyanate 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Cyanophenylisocyanate, 3-Isocyanatobenzonitrile
314180050 314180010
ACRO314180050EA 1775 SEK
ACRO314180050 ACRO314180010
3-Cyanophenyl isocyanate 97%
3-Cyanophenyl isocyanate
Formel: C₈H₄N₂O
MW: 144,13 g/mol
Kokpunkt: 123 °C (12 mmHg)
Smältpunkt: 49…54 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013856
CAS nummer: 16413-26-6
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Off-white to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥96.0 %