3,5-Bis(trifluoromethyl)benzaldehyde 97%
Leverantör: Thermo Fisher Scientific
Varning
307690050 307690010 307690250
ACRO307690050EA 1675 SEK
ACRO307690050 ACRO307690010 ACRO307690250
3,5-Bis(trifluoromethyl)benzaldehyde 97%
3,5-Bis(trifluoromethyl)benzaldehyde
Formel: C₉H₄F₆O
MW: 242.12 g/mol
Kokpunkt: 37 °C (1.3 mmHg)
Smältpunkt: -2
Densitet: 1.47 g/cm³
Flampunkt: 69 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010206
CAS nummer: 401-95-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4215 to 1.4235 (20°C, 589 nm)
GC ≥96.0 %