4-tert-Butylbenzenesulphonyl chloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-tert-Butylbenzene-1-sulphonyl chloride
303470100 303470500
ACRO303470100EA 970 SEK
ACRO303470100 ACRO303470500
4-tert-Butylbenzenesulphonyl chloride 98%
4-tert-Butylbenzenesulphonyl chloride
Formel: C₁₀H₁₃ClO₂S
MW: 232,73 g/mol
Kokpunkt: 165 °C (18 mmHg)
Smältpunkt: 79…83 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007449
CAS nummer: 15084-51-2
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to off-white to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 78 to 84 °C
GC ≥97.5 %