5-Phenyl-1H-tetrazole 99%
Leverantör: Thermo Scientific
Varning
263720050 263720250 263721000
ACRO263720050EA 370 SEK
ACRO263720050 ACRO263720250 ACRO263721000
5-Phenyl-1H-tetrazole 99%
5-Phenyl-1H-tetrazole
Formel: C₇H₆N₄
MW: 146,15 g/mol
Kokpunkt: 331 °C (1013 hPa)
Smältpunkt: 222 °C
Storage Temperature: Omgivande
MDL Number: MFCD00022388
CAS nummer: 18039-42-4
EINECS: 241-950-8
UN: 1325
ADR: 4.1,II
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %