1-Amino-3-methoxypropane 99+%
Leverantör: Thermo Fisher Scientific
Fara
257520010 257520050
ACRO257520010EA 1075 SEK
ACRO257520010 ACRO257520050
1-Amino-3-methoxypropane 99+%
1-Amino-3-methoxypropane
Formel: C₄H₁₁NO
Kokpunkt: 118 °C (1013 hPa)
Smältpunkt: –65 °C
Densitet: 0,87 g/cm³
Flampunkt: 48 °C
MDL Number: MFCD00014831
CAS nummer: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,I
Order Now
Specifikation testresultat
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4185 (20°C, 589 nm)
GC ≥99.0 %