5-Bromouracil 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil
226440050 226440250
ACRO226440050EA 475 SEK
ACRO226440050 ACRO226440250
5-Bromouracil 98%
5-Bromouracil
Formel: C₄H₃BrN₂O₂
MW: 190,98 g/mol
Kokpunkt: 384 °C (1013 hPa)
Smältpunkt: 307…310 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006017
CAS nummer: 51-20-7
EINECS: 200-084-0
Merck Index: 13,01425
Order Now
Specifikation testresultat
Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %