2-Chloroethyl methyl ether 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Methoxyethyl chloride
221771000 221775000 221770025
ACRO221771000EA 1175 SEK
ACRO221771000 ACRO221775000 ACRO221770025
2-Chloroethyl methyl ether 98%
2-Chloroethyl methyl ether
Formel: C₃H₇ClO
MW: 94,54 g/mol
Kokpunkt: 89…90 °C (760 mmHg)
Smältpunkt: –55 °C
Densitet: 1,03 g/cm³
Flampunkt: 15 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000972
CAS nummer: 627-42-9
EINECS: 210-999-7
UN: 1992
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4100 (20°C, 589 nm)
GC ≥97.5 %