Octyltrichlorosilane 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Trichloro(octyl)silane
216501000 216505000
ACRO216501000EA 1025 SEK
ACRO216501000 ACRO216505000
Octyltrichlorosilane 97%
Octyltrichlorosilane
Formel: C₈H₁₇Cl₃Si
MW: 247.67 g/mol
Kokpunkt: 233 °C (731 mmHg)
Densitet: 1.07 g/cm³
Flampunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000488
CAS nummer: 5283-66-9
EINECS: 226-112-1
UN: 1801
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20°C, 589 nm)
GC ≥96.0 %