(α,α,α-Trifluoro-4-tolyl)acetic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (4-(Trifluoromethyl)phenyl)acetic acid, 4-(Trifluoromethyl)phenylacetic acid
214370010 214370050
ACRO214370010EA 635 SEK
ACRO214370010 ACRO214370050
(α,α,α-Trifluoro-4-tolyl)acetic acid 98%
(α,α,α-Trifluoro-4-tolyl)acetic acid
Formel: (C₆H₄(CF₃))CH₂COOH
MW: 204.15 g/mol
Smältpunkt: 81…85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004352
CAS nummer: 32857-62-8
EINECS: 251-263-5
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 81 to 85 °C
Titration with NaOH ≥97.5 %