Benzyltrimethylammonium chloride 98+%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride
211862500 211860010 211860500
ACRO211862500EA 795 SEK
ACRO211862500 ACRO211860010 ACRO211860500
Benzyltrimethylammonium chloride 98+%
Benzyltrimethylammonium chloride
Formel: C₁₀H₁₆ClN
Smältpunkt: 239 °C (dec.)
Densitet: 1,07 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011782
CAS nummer: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %