Benzyl isocyanate 99+%
Leverantör: Thermo Fisher Scientific
Fara
210760050 210760250
ACRO210760050EA 1150 SEK
ACRO210760050 ACRO210760250
Benzyl isocyanate 99+%
Benzyl isocyanate
Formel: C₈H₇NO
MW: 133.15 g/mol
Kokpunkt: 101…104 °C (33 mmHg)
Densitet: 1.07 g/cm³
Flampunkt: 45
MDL Number: MFCD00009701
CAS nummer: 3173-56-6
EINECS: 221-640-9
UN: 3080
ADR: 6.1 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5241 to 1.5261 (20°C, 589 nm)
GC ≥99.0 %