Vinyltrimethylsilane 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Trimethyl(vinyl)silane
200330250 200331000
ACRO200330250EA 1250 SEK
ACRO200330250 ACRO200331000
Vinyltrimethylsilane 97%
Vinyltrimethylsilane
Formel: C₅H₁₂Si
MW: 100.24 g/mol
Kokpunkt: 55
Densitet: 0.68 g/cm³
Flampunkt: –24 °C
MDL Number: MFCD00008606
CAS nummer: 754-05-2
EINECS: 212-042-9
UN: 1993
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3895 to 1.3915 (20°C, 589 nm)
GC ≥96.0 %