Tris(triphenylphosphine)rhodium(I) chloride 99.99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium
194570010 194570050 194572500
ACRO194570010EA 5475 SEK
ACRO194570010 ACRO194570050 ACRO194572500
Tris(triphenylphosphine)rhodium(I) chloride 99.99%
Tris(triphenylphosphine)rhodium(I) chloride
Formel: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Smältpunkt: 245…250 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010016
CAS nummer: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226
Order Now
Specifikation testresultat
Appearance Brown Crystalline powder
Infrared spectrum Conforms
Total trace metal impurities ≤0.01 %