Thiophene-3-carbaldehyde 98%
Leverantör: Thermo Fisher Scientific
Varning
187980050 187980250 187981000
ACRO187980050EA 1475 SEK
ACRO187980050 ACRO187980250 ACRO187981000
Thiophene-3-carbaldehyde 98%
Thiophene-3-carbaldehyde
Formel: C₅H₄OS
Kokpunkt: 194…196 °C (760 mmHg)
Smältpunkt: –30 °C
Densitet: 1,28 g/cm³
Flampunkt: 64 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005466
CAS nummer: 498-62-4
EINECS: 207-865-5
Order Now
Specifikation testresultat
Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5860 (20°C, 589 nm)
GC ≥97.5 %