m-Fenetedin 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Etoxianilin
186720250 186720050
ACRO186720250EA 2150 SEK
ACRO186720250 ACRO186720050
m-Fenetedin 98%
m-Fenetedin
Formel: C₈H₁₁NO
MW: 137,18 g/mol
Kokpunkt: 248 °C (1013 hPa)
Densitet: 1,03 g/cm³
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007785
CAS nummer: 621-33-0
EINECS: 210-680-2
UN: 2311
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5670 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %