n-Butyllithium 1.6 M hexanes, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
181271000 181275000 181278000
181271000.EA 1100 SEK
181271000. 181275000. 181278000.
n-Butyllithium 1.6 M hexanes, AcroSeal®
n-Butyllithium
Formel: C₄H₉Li
MW: 64.06 g/mol
Smältpunkt: –95 °C
Densitet: 0.68 g/cm³
Flampunkt: –18 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009414
CAS nummer: 109-72-8
EINECS: 203-698-7
UN: 3394
ADR: 4.2 I
Order Now
Specifikation testresultat
Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.49 to 1.70 M
Free base ≤1 %