Etylklorformiat 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Chloroformic acid ethyl ester
181080050 181085000 181080025 181081000
ACRO181080050EA 630 SEK
ACRO181080050 ACRO181085000 ACRO181080025 ACRO181081000
Etylklorformiat 97%
Etylklorformiat
Formel: C₃H₅ClO₂
MW: 108,52 g/mol
Kokpunkt: 94 °C (760 mmHg)
Smältpunkt: –81 °C
Densitet: 1,135 g/cm³
Flampunkt: 10 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000644
CAS nummer: 541-41-3
EINECS: 208-778-5
UN: 1182
ADR: 6.1,
Merck Index: 12,03831
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤100 APHA