Phenoxyacetic acid 98+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Phenoxyacetic acid
169870050 169871000 169875000 169870025
ACRO169870050EA 350 SEK
ACRO169870050 ACRO169871000 ACRO169875000 ACRO169870025
Phenoxyacetic acid 98+%
Phenoxyacetic acid
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 285 °C (760 mmHg)
Smältpunkt: 98…101 °C
Densitet: 1,36 g/cm³ (20 °C)
Flampunkt: 165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004296
CAS nummer: 122-59-8
EINECS: 204-556-7
Merck Index: 12,07407
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Titration with NaOH ≥98.0 %
NMR (1H-NMR) Consistent with structure