Metakrylsyra 99.5% Stabilised, extra_pure
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Metylpropensyra
168310010 168310040 168310250 168312500 168315000 168310251
ACRO168310010EA 1625 SEK
ACRO168310010 ACRO168310040 ACRO168310250 ACRO168312500 ACRO168315000 ACRO168310251
Metakrylsyra 99.5% Stabilised, extra_pure
Metakrylsyra
Formel: C₄H₆O₂
MW: 86.09 g/mol
Kokpunkt: 163 °C (760 mmHg)
Smältpunkt: 16 °C
Densitet: 1.01 g/cm³
Flampunkt: 65 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002651
CAS nummer: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8 II
Merck Index: 13,05967
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4295 to 1.4315 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.4 %
Color scale ≤20 APHA
Stabilizer ca. 250 ppm MEHQ