Valeronitrile 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Pentanenitrile
167941000 167945000
ACRO167941000EA 905 SEK
ACRO167941000 ACRO167945000
Valeronitrile 98%
Valeronitrile
Formel: C₅H₉N
MW: 83.13 g/mol
Kokpunkt: 139…141 °C (760 mmHg)
Smältpunkt: –96 °C
Densitet: 0.79 g/cm³
Flampunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001974
CAS nummer: 110-59-8
EINECS: 203-781-8
UN: 1992
ADR: 3 III
Merck Index: 12,10043
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589nm)
GC ≥97.5 %