Ethyl crotonate 96%
Leverantör: Thermo Scientific
Varning
Synonyms: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester
161381000 161385000
ACRO161381000EA 605 SEK
ACRO161381000 ACRO161385000
Ethyl crotonate 96%
Ethyl crotonate
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 143 °C (1013 hPa)
Smältpunkt: –79 °C
Densitet: 0,918 g/cm³
Flampunkt: 34 °C
MDL Number: MFCD00009289
CAS nummer: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4280 (20°C, 589 nm)
Density (20/4°C) 0.916 to 0.921 g/ml
GC ≥95 %
Acid value ≤1 mg KOH/g