tert-Butyl bromoacetate 99%
Leverantör: Thermo Fisher Scientific
Varning
154340100 154340500 154342500
ACRO154340100EA 755 SEK
ACRO154340100 ACRO154340500 ACRO154342500
tert-Butyl bromoacetate 99%
tert-Butyl bromoacetate
Formel: BrCH₂COOC(CH₃)₂
Kokpunkt: 50 °C (10 mmHg)
Densitet: 1,338 g/cm³
Flampunkt: 49 °C
MDL Number: MFCD00000188
CAS nummer: 5292-43-3
EINECS: 226-133-6
UN: 1993
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure