Benzothiazole 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Benzo[d]thiazole
146390010 146390050 146392500 146390025
ACRO146390010EA 3250 SEK
ACRO146390010 ACRO146390050 ACRO146392500 ACRO146390025
Benzothiazole 97%
Benzothiazole
Formel: C₇H₅NS
MW: 135.19 g/mol
Kokpunkt: 231 °C (760 mmHg)
Smältpunkt: 2 °C
Densitet: 1.23 g/cm³
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005775
CAS nummer: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: 6.1 III
Merck Index: 13,01108
Order Now
Specifikation testresultat
Appearance Clear yellow-brown to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6410 to 1.6440 (20°C, 589 nm)
GC ≥96.0 %