Trimethylsulphonium iodide 98%
Leverantör: Thermo Scientific
Varning
140250250 140251000 140255000
ACRO140250250EA 610 SEK
ACRO140250250 ACRO140251000 ACRO140255000
Trimethylsulphonium iodide 98%
Trimethylsulphonium iodide
Formel: C₃H₉IS
MW: 204,07 g/mol
Smältpunkt: 215…220 °C (subl.)
Densitet: 1,922 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011632
CAS nummer: 2181-42-2
EINECS: 218-555-4
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥96.0 %