p-Tolylacetic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(p-Tolyl)acetic acid, 4-Tolylacetic acid, (4-Methylphenyl)acetic acid
139120250 139120050 139121000
ACRO139120250EA 915 SEK
ACRO139120250 ACRO139120050 ACRO139121000
p-Tolylacetic acid 99%
p-Tolylacetic acid
Formel: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Kokpunkt: 265…267 °C (760 mmHg)
Smältpunkt: 90…93 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004353
CAS nummer: 622-47-9
EINECS: 210-738-7
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 93 °C
Titration with NaOH ≥98.5 %