2-Thiophenemethanol 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
138860250 138861000
ACRO138860250EA 935 SEK
ACRO138860250 ACRO138861000
2-Thiophenemethanol 99+%
2-Thiophenemethanol
Formel: C₅H₆OS
MW: 114,17 g/mol
Kokpunkt: 206…207 °C (761 mmHg)
Smältpunkt: 0 °C
Densitet: 0,9 g/cm³
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005454
CAS nummer: 636-72-6
EINECS: 211-264-3
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5660 (20°C, 589 nm)
GC ≥99.0 %