Ftalimid 99%
Leverantör: Thermo Scientific
Synonyms: Phthalic acid imide
131100010 131101000 131105000 131100025
ACRO131100010EA 830 SEK
ACRO131100010 ACRO131101000 ACRO131105000 ACRO131100025
Ftalimid 99%
Ftalimid
Formel: C₈H₅NO₂
MW: 147,13 g/mol
Kokpunkt: 366 °C (760 mmHg)
Smältpunkt: >233,5 °C
Densitet: 1,21 g/cm³ (20 °C)
Flampunkt: 165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005881
CAS nummer: 85-41-6
EINECS: 201-603-3
Merck Index: 12,07529
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or flakes
Infrared spectrum Conforms
Melting point 232 to 236 °C
HPLC ≥98.5 %