N-Methylmaleimide 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Methyl-1H-pyrrole-2,5-dione
127080050 127080250 127081000
ACRO127080050EA 1125 SEK
ACRO127080050 ACRO127080250 ACRO127081000
N-Methylmaleimide 97%
N-Methylmaleimide
Formel: C₅H₅NO₂
MW: 111,1 g/mol
Kokpunkt: 194 °C (1013 hPa)
Smältpunkt: 93…98 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005508
CAS nummer: 930-88-1
EINECS: 213-226-1
UN: 2928
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Light yellow Crystals
Infrared spectrum Conforms
Melting point 93 to 98 °C
GC ≥96.0 %