Methyl-2,4-dihydroxybenzoate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,4-Dihydroxybenzoic acid methyl ester, Methyl 2,4-dihydroxybenzoate
126730250 126730050 126731000
ACRO126730250EA 1650 SEK
ACRO126730250 ACRO126730050 ACRO126731000
Methyl-2,4-dihydroxybenzoate 97%
Methyl-2,4-dihydroxybenzoate
Formel: C₈H₈O₄
MW: 168,15 g/mol
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 116…121 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002276
CAS nummer: 2150-47-2
EINECS: 218-428-3
Order Now
Specifikation testresultat
Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 116 to 121 °C
GC ≥96.0 %
HPLC ≥97.5 %